missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX HitHunter™ cAMP Assay for Biologics

Product Code. 17322339
Encompass
Change view
Click to view available options
Quantity:
1,000/4,000 dp (96-well/384-well)
Unit Size:
1 set
Product Code. Quantity unitSize
17322339 1,000/4,000 dp (96-well/384-well) 1 set
1 options
This item is not returnable. View return policy

Product Code. 17322339

Brand: DiscoverX 900075LM10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The HitHunter™ cAMP Assay for Biologics is an EFC-based detection kit to assay levels of cellular cAMP. This detection kit contains reagents for assaying 10 plates (96-well).

This detection kit is a robust, sensitive and easy-to-use assay to determine GPCR activity through detection of intracellular cAMP levels. The HitHunter cAMP assay is intended for use with cells expressing Gs or Gi coupled GPCRs. This product uses EFC technology, and contains ED-tagged cAMP, anti-cAMP antibody, and pure EA. Upon application to a cell which contains endogenous, untagged cAMP, the anti-cAMP antibody will be sequestered by binding to this untagged cAMP, allowing for EFC between the tagged cAMP and soluble EA. This will produce a proportional level of chemiluminescent signal. This kit format is intended for Biologics, including crude preps, antibodies, and more, and is written for a 96-well plate. This kit may be used with Eurofins DiscoverX cAMP cell lines as well as cell lines from other sources. This detection kit contains reagents for assaying 10 plates (96-well).

Specifications

Assay Entry GPCR cAMP
For Use With (Application) Detection Kits
Quantity 1,000/4,000 dp (96-well/384-well)
Target Entry cAMP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.