Learn More
Histone Deacetylase Inhibitor II, Thermo Scientific™
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Quantity | 5 mg |
| Description | Off-white |
| Content And Storage | -30°C to -10°C |
| CAS | 174664-65-4 |
| Molecular Formula | C10H10N2O4 |
| Formula Weight | 222.20g/mol |
| Synonym | 3-Carboxy cinnamic acid bis-hydroxamide; CBHA |
| Sensitivity | Light Sensitive |
| Product Type | Signal Transduction Reagent |
| Form | Powder |