missing translation for 'onlineSavingsMsg'
Learn More

Histone Acetyltransferase Inhibitor II, MedChemExpress

Product Code. p-200099862
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. Quantity unitSize
16493138 10 mg 10mg
16482636 25 mg 25mg
16439696 50 mg 50mg
16435877 5 mg 5mg
16424436 100 mg 100mg
5 options
This item is not returnable. View return policy

Product Code. 16493138

Brand: Medchem Express HY100734

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Histone Acetyltransferase Inhibitor II (compound 2c) is a potent, selective and cell permeable p300 histone acetyltransferase inhibitor, with an IC50 of 5 μM. Histone Acetyltransferase Inhibitor II shows anti-acetylase activity in mammalian cells.

Histone Acetyltransferase Inhibitor II (compound 2c) is a potent, selective and cell permeable p300 histone acetyltransferase inhibitor, with an IC50 of 5 μM. Histone Acetyltransferase Inhibitor II shows anti-acetylase activity in mammalian cells.

Specifications

Chemical Name or Material Histone Acetyltransferase Inhibitor II
CAS 932749-62-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H16Br2O3
Quantity 10 mg
Solubility Information DMSO : ≥ 30 mg/mL (64.63 mM)
SMILES O=C1/C(CCC/C1=C\C2=CC=C(O)C(Br)=C2)=C/C3=CC=C(O)C(Br)=C3
Molecular Weight (g/mol) 464.15
Formula Weight 464.15
Percent Purity 98.72%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.