Learn More
HIST1H4F Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18341-1-AP-150UL
This item is not returnable.
View return policy
Specifications
| HIST1H4F | |
| Polyclonal | |
| Unconjugated | |
| HIST1H4F | |
| H4, H4/A, H4/B, H4/c, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, HIST4H4, histone cluster 1, H4f, Histone H4, histoneh4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 319157, 8361 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P62805 | |
| Hist1h4f | |
| HIST1H4F Fusion Protein Ag13113 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |