Learn More
HIST1H4F Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. 16858424 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16858424 20 μL 20µL
16848424 150 μL 150µL
2 options
Product Code. 16858424
Brand: Proteintech 183411AP20UL
Rabbit Polyclonal Antibody
Specifications
Antigen HIST1H4F
Applications Western Blot
Classification Polyclonal
Concentration 0.39 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HIST1H4F
Gene Accession No. P62805
Gene Alias H4, H4/A, H4/B, H4/c, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, HIST4H4, histone cluster 1, H4f, Histone H4, histoneh4
Gene Symbols Hist1h4f
Host Species Rabbit
Immunogen HIST1H4F Fusion Protein Ag13113
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 319157, 8361
Target Species Rat, Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG