missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIST1H1T Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18188-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a replication-dependent histone that is a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element.Specifications
HIST1H1T | |
Polyclonal | |
Unconjugated | |
HIST1H1T | |
H1FT, H1t, HIST1H1T, histone cluster 1, H1t, Histone H1t, Testicular H1 histone | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
107970, 3010 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P22492, Q07133 | |
Hist1h1t | |
HIST1H1T Fusion Protein Ag12908 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |