Learn More
Hirschmann™ Storage Bottle
Product Code. 10773321
Change view
Click to view available options
Capacity (Metric):
1000 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10773321 1000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10773321 Supplier Hirschmann™ Supplier No. 8901793
  • ONLINE EXCLUSIVE PRICE
    279.30 NOK
    222.28 NOK / Pack of 1
    Save 57.02 NOK
    20% Off
Estimated Shipment: 08-09-2026
Specifications
Capacity (Metric) 1000 mL
Material Glass
Product Type Storage Bottle
For Use With (Equipment) Automatic Pipettes