missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syngene™ HILED Lighting Option
Shop All Syngene Products

Click to view available options
Color:
Green, Blue, Far Red
Green, Blue, Infa Red, Far Red
Infa Red, Far Red
Red, Blue, Green
Red, Blue, Green, InfraRed
Red, InfraRed
Unit Size:
Each
Description
- For use with GBOX Mini only
- Including gantry, electronics, and lighting units
Specifications
Specifications
| Color | Red, InfraRed |
| For Use With (Equipment) | GBOX Mini |
| Includes | Gantry, electronics, lighting units |
| Product Type | Lighting |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction