missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hikeshi Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20524-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Required for organization and/or function of the secretory apparatus in Clara cells in lung.Specifications
Hikeshi | |
Polyclonal | |
Unconjugated | |
HIKESHI | |
C11orf73, Hikeshi, HSPC138, HSPC179, HSPC248, L7RN6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
293103, 51501, 67669 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q53FT3, Q5M808, Q9DD02 | |
HIKESHI | |
Hikeshi Fusion Protein Ag14376 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |