Learn More
Thermo Scientific™ HIGHPlate 6000 Microplate Rotor for 4 L General Purpose X Pro Centrifuges
Product Code. 10227814 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity:
2 x 5 Standard Plates
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
10227814 2 x 5 Standard Plates Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10227814 Supplier Thermo Scientific™ Supplier No. 75003606
  • ONLINE EXCLUSIVE PRICE
    52760.00 NOK
    50538.73 NOK / Pack of 1
    Save 2221.27 NOK
    4% Off
 Restricted Item
In Stock
Add to basket
Add to basket
Spin filter plates with the Thermo Scientific™ HIGHPlate™ 6000 Microplate Rotor.
Spin filter plates with the Thermo Scientific™ HIGHPlate™ 6000 Microplate Rotor, for use with DNA purification protocols.
Compatible with:
Thermo Scientific™ Sorvall™ Legend™ XT/XTR, Multifuge™ X3/X3R
TRUSTED_SUSTAINABILITY
Specifications
Description HIGHPlate 6000 Microplate Rotor
Type Swinging Bucket Rotor
Capacity 2 x 5 Standard Plates
Max. RCF 6,168 x g
Max. Speed 6,300 rpm
Radius (Metric) 139 mm
For Use With (Equipment) Thermo Scientific Sorvall Legend XT/XF, Multifuge X3/X3F and SL 40F Centrifuges
Angle 90°
Certifications/Compliance Biocontainment certified by CAMR™, HPA, Porton Down, U.K.
Biocontainment Yes