Learn More
Hamilton™ 1000μL Conductive Non-Filter, Non-Sterile CO-RE Pipet Tips
Description
- 95.1mm tip length
- Color code label: white
- Usable with 1000μL pipetting channels/CO-RE 96 Probe Head
- Pressure-based Liquid Level Detection (pLLD) and capacitive (cLLD) with conductive (black) version
- Process safety offered by Monitored Air Displacement and Total Aspiration and Dispense Monitoring
Specifications
Specifications
| Color | Black |
| Volume (Metric) | 1000 μL |
| Sterility | Non-sterile |