Learn More
Velp Scientifica™ High Temperature Sealing Grease
Product Code. 15399604 Shop All Velp Products
Change view
Click to view available options
For Use With:
Dumas Nitrogen Analyzer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With Unit Size
15399604 Dumas Nitrogen Analyzer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15399604 Supplier Velp Scientifica™ Supplier No. A00000236
  • ONLINE EXCLUSIVE PRICE
    181.36 NOK
    169.63 NOK / Pack of 1
    Save 11.73 NOK
    6% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Velp Scientifica™ High Temperature Sealing Grease for use with NDA 701 /702 and OXITEST.
  • For Use With (Equipment): NDA 701 /702 and OXITEST
Specifications
Type High Temperature Sealing Grease
For Use With Dumas Nitrogen Analyzer
Weight (Metric) 0.02 kg