Learn More
Nabertherm™ High-Temperature Ovens, Forced Convection Chamber Furnaces
Product Code. 17368059 Shop All Nabertherm Products
Change view
Click to view available options
Capacity (Metric):
30 L
60 L
120 L
250 L
500 L
Controller Type:
B400
B500
B510
C540
C550
Unit Size:
Each
Product Code. Capacity (Metric) Controller Type unitSize
17368059 250 L C540 Each
17338049 60 L B500 Each
17348049 60 L C540 Each
17398049 120 L B500 Each
17308059 120 L C540 Each
17358059 250 L B400 Each
17318069 500 L B400 Each
17328069 500 L C540 Each
17378069 30 L B510 Each
17388069 30 L C550 Each
10 options
Product Code. 17368059
Brand: Nabertherm™ NA2583M2
Only null left
Add to basket
Add to basket
Chamber furnaces with air circulation are characterized by their extremely high temperature uniformity. Suitable for processes such as annealing, crystalizing, preheating, curing, and tool making.
  • Horizontal air circulation with optimum distribution through stainless steel baffles
  • Swing door hinged on the right
  • Temperature uniformity up to +/- 4 C according to DIN 17052-1 (NAT 15/65 and NAT 30/85 up to +/- 5°C) in the empty work space
  • One frame sheet and rails for two additional trays included in the scope of delivery (NAT 15/65 and NAT 30/85 without frame sheet)
  • Base frame included in the delivery, NAT 15/65 and NAT 30/85 designed as table-top model
Specifications
Controller Type C540
Capacity (Metric) 250 L
Temperature (Metric) 850°C
Dimensions (D x W x H) Interior 750 x 600 x 600 mm
Dimensions (L x W x H) Exterior 1690 x 1120 x 1810 mm
Voltage 400 V
Wattage 23000 W