Learn More
Nabertherm™ High-Temperature Furnaces with Scale for Determination of Combustion Loss and Thermogravimetric Analysis (TGA)
Product Code. 17397829 Shop All Nabertherm Products
Change view
Click to view available options
Capacity (Metric):
16 L
17 L
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
17397829 16 L Each
17307839 17 L Each
2 options
Product Code. 17397829
Brand: Nabertherm™ 001711560
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
These high-temperature furnaces are specially developed to determine combustion loss during annealing and for thermogravimetric analysis in the lab. The complete system consists of the high-temperature furnace, a table frame, precision scale with feedthroughs into the furnace and powerful software.
  • High-quality molybdenum disilicide heating elements
  • Dual shell housing made of textured stainless steel sheets with additional fan cooling for low surface temperature
  • Adjustable air inlet
  • Exhaust air opening in the roof
  • 4 scales available for different maximum weights and scaling ranges
  • Process control and documentation for temperature and combustion loss via VCD software package for monitoring
Specifications
Controller Type P570
Capacity (Metric) 16 L
Temperature (Metric) 1600°C
Dimensions (D x W x H) Interior 150 x 150 x 150 mm
Dimensions (L x W x H) Exterior 370 x 655 x 890 mm
Voltage 400 V
Wattage 5000 W