Learn More
Nabertherm™ High-Temperature Bottom Loading Furnaces
The motor-driven lifting table significantly simplifies the charging of the high-temperature furnaces. The heating all around the cylindrical furnace chamber provides for an optimal temperature uniformity. Up to 3 charge saggars can be stacked on top of each other resulting in a high productivity.
Brand:  Nabertherm™ LHB157WN3
Additional Details : Weight : 43.00000kg
Product Code. 17367979
  • 123536.39 NOK / Each
Estimated Shipment: 04-06-2024
Description
  • High-quality heating elements made of molybdenum disilicide offer very good protection against chemical interaction between charge and heating elements
  • Excellent temperature uniformity thanks to three (LHT 02/17 LB Speed) or four-sided (LHT 01/17 LB Speed) heating of the furnace chamber
  • Precise, motorized toothed belt drive of the table with button operation
  • Exhaust air vent in the roof
Specifications
Specifications
P580
1650°C
590 x 350 x 680 mm
2900 W
1 L
180 x 145 x 100 mm
230 V