Learn More
Cytiva High Purity dNTPs
Product Code. 11515115 Shop All Cytiva Products
Change view
Click to view available options
Product Type:
dATP
dCTP
dGTP
dTTP
Unit Size:
250mg
Product Code. Product Type unitSize
11515115 dCTP 250mg
11505115 dATP 250mg
11525115 dGTP 250mg
11535115 dTTP 250mg
4 options
Product Code. 11515115
Brand: Cytiva 27186004
Used for amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis, and expression profiling. Cytiva High Purity dNTPs are tested functionally in long PCR* to be PCR-quantified for high-quality sequencing.
  • High purity: >98% triphosphate, as confirmed by chromatographic analysis and 31P nuclear magnetic resonance (NMR) spectroscopy
  • Supplied as solids and also available in solutions
TRUSTED_SUSTAINABILITY
Specifications
Format Solid
Product Type dCTP
Content And Storage -20°C
Purity ≥98% triphosphate
Quantity 250 mg
For Use With (Application) Amplification, Dideoxy sequencing, labeling, mutagenesis, cDNA synthesis and expression profiling