Learn More
Greiner Bio-One High Profile Polystyrene Microplate Lids
GREENER_CHOICE
Product Code. 10375382
Click to view available options
Sterility:
Non-sterile
Condensation Rings:
Yes
No. per Pack:
1
No. per Case:
100
Unit Size:
Pack of 100
Pack of 400
Product Code. 10375382
Brand: Greiner Bio-One 656170
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For use with non TC-treated 96-well microplates. Greiner Bio-one High Profile Polystyrene Microplate Lids is free of detectable DNase, RNase, human DNA.
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic, non-cytotoxic
  • Offer protection against contamination and evaporation during sample storage and cell cultivation
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Dimensions (L x W x H) 128 x 85 x 10 mm
Material Polystyrene
Sterility Non-sterile
Condensation Rings Yes
No. per Case 100
No. per Pack 1
Evaporation Barrier Yes
For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.