Learn More
Scotch-Brite™ High Performance Cleaning Wipes 2011
Description
- For removal of dust, dirt, grease, and fingerprints
- Use on stainless steel, chrome, plastics, glass, paint lacquers, and other finished or polished metal surfaces
- Eliminates the need for cleaning agents in many cases
- Machine washable up to 75 times at 95°C
Specifications
Specifications
| Product Type | Wipes |
| Length (Metric) | 360 mm |
| Width (Metric) | 320 mm |
| Color | Multiple |
| Dimensions (L x W) | 360 x 320 mm |
| Packaging Quantity | 3/Pk. |
| Care Instructions | Machine Washable |