Learn More
Kartell™ High Impact Polystyrene Balance Boats
Product Code. 12627805
Change view
Click to view available options
Capacity (Metric):
5 mL
7 mL
30 mL
100 mL
250 mL
Color:
Black
White
Shape:
Diamond
Square
Unit Size:
Pack of 1000
Pack of 250
Pack of 500
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Color Shape Unit Size
12627805 7 mL White Square Pack of 1000
10447512 5 mL White Diamond Pack of 1000
10325962 5 mL Black Diamond Pack of 1000
10662163 30 mL White Diamond Pack of 500
10192122 30 mL Black Diamond Pack of 500
10738294 100 mL White Diamond Pack of 250
10345812 100 mL Black Diamond Pack of 250
10736962 100 mL White Square Pack of 1000
10325781 250 mL White Square Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 12627805 Supplier Kartell™ Supplier No. 1110300
  • ONLINE EXCLUSIVE PRICE
    950.00 NOK
    930.45 NOK / Pack of 1000
    Save 19.55 NOK
    2% Off
View all promotions
 Restricted Item
In Stock
Provide an easy and inexpensive system for weighing any medium.
  • Lightweight weighing boats
  • Non-contaminating (made of HIPS with hydrophobic surface)
  • Easily fashioned to make funnel shape
  • Available in two models: diamond (DS) and square (SQ) 
  • DS model available in white or black for visibility of trace sample remains adhering to boat
  • Reproducible tare weight: batch to batch variation less than 5%
  • Antistatic
Specifications
Bottom Shape Flat
Capacity (Metric) 7 mL
Color White
Material Polystyrene
Shape Square
Type Weighing Boat
Dimensions (L x W x H) 43 x 43 x 8 mm
Length (Metric) 43 mm
Width (Metric) 43 mm
Height (Metric) 8 mm
Weight (Metric) 0.5 g