missing translation for 'onlineSavingsMsg'
Learn More

HIC2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 22788-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16892085

  • 4593.76 NOK / 150µL
Estimated Shipment: 13-08-2024
to see stock.

Description

Description

HIC2 contains 5 C2H2-type zinc fingers and 1 BTB (POZ) domain. It belongs to the krueppel C2H2-type zinc-finger protein family, Hic subfamily and is a transcriptional repressor.
Specifications

Specifications

HIC2
Polyclonal
Unconjugated
HIC2
Hic 2, Hic 3, HIC2, HRG22, hypermethylated in cancer 2, KIAA1020, ZBTB30
Rabbit
Antigen Affinity Chromatography
RUO
23119
-20°C
Liquid
Immunoprecipitation, Western Blot
0.61 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q96JB3
HIC2
HIC2 Fusion Protein Ag18789
150 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers