Learn More
Erlab HF4 U16
Product Code. 16832903
Change view
Click to view available options
For Use With (Equipment):
Fume Hood
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
16832903 Fume Hood Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16832903 Supplier Erlab Supplier No. H11014061
Filter for your air purifier Halo 35 P.
  • Filter for your air purifier 35 P
  • Type of filter: Particulate (ULPA)
  • Dimensions: 757,5 x 406,5 x 89 mm
  • Quantity/Set: 1
Specifications
For Use With (Equipment) Fume Hood
Product Type HEPA Filter