Learn More
Hexythiazox, TRC
Product Code. 30132604
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132604 10 g 10g
30138588 1 g 1g
17971245 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30132604 Supplier Toronto Research Chemicals Supplier No. TRCH29850010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 78587-05-0
Molecular Formula C17 H21 Cl N2 O2 S
Molecular Weight (g/mol) 352.88
Synonym 3-Thiazolidinecarboxamide, 5-(4-chlorophenyl)-N-cyclohexyl-4-methyl-2-oxo-, (4R,5R)-rel-, 3-Thiazolidinecarboxamide, 5-(4-chlorophenyl)-N-cyclohexyl-4-methyl-2-oxo-, trans-, Calibre, Cesar, GWN 2106, Hexygon, Hexythiazox, Matacar, NA 73, Nissorun, Onager, Ordoval, Savey, Savey PM, Trevi, Zeldox
IUPAC Name (4S,5S)-5-(4-chlorophenyl)-N-cyclohexyl-4-methyl-2-oxo-1,3-thiazolidine-3-carboxamide
SMILES C[C@H]1[C@@H](SC(=O)N1C(=O)NC2CCCCC2)c3ccc(Cl)cc3
Specifications
Quantity 10 g
InChI Formula InChI=1S/C17H21ClN2O2S/c1-11-15(12-7-9-13(18)10-8-12)23-17(22)20(11)16(21)19-14-5-3-2-4-6-14/h7-11,14-15H,2-6H2,1H3,(H,19,21)/t11-,15+/m0/s1
Formula Weight 352.1012
Chemical Name or Material Hexythiazox