Learn More
HEXIM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16598-1-AP-20UL
This item is not returnable.
View return policy
Description
Transcriptional regulator which functions as a general RNA polymerase II transcription inhibitor. In cooperation with 7SK snRNA sequesters P-TEFb in a large inactive 7SK snRNP complex preventing RNA polymerase II phosphorylation and subsequent transcriptional elongation.Specifications
| HEXIM2 | |
| Polyclonal | |
| Unconjugated | |
| HEXIM2 | |
| HEXIM2, L3, Protein HEXIM2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 124790, 303580, 71059 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q3TVI4, Q96MH2 | |
| Hexim2 | |
| HEXIM2 Fusion Protein Ag9754 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |