Learn More
Hexestrol, TRC
Product Code. 30393597
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30393597 500 mg
30389132 100 mg
30401848 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30393597 Supplier Toronto Research Chemicals Supplier No. TRCH295300500MG
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 84-16-2
Molecular Formula C18 H22 O2
Molecular Weight (g/mol) 270.37
Synonym Phenol, 4,4'-(1,2-diethyl-1,2-ethanediyl)bis-, (R*,S*)-, Phenol, 4,4'-(1,2-diethylethylene)di-, meso- (8CI), Cycloestrol, Erythrohexestrol, Estrifar, Estronal, Extra-Plex, Hexanoestrol, Hexestrofen, Hexestrol, Hexoestrol, Hexron, Hormoestrol, Mesohexestrol, NSC 9894, Sinestrol, Synestrol, Synoestrol, Synthovo, Syntrogene, meso-3,4-Bis(p-hydroxyphenyl)-n-hexane, meso-3,4-Di(p-hydroxyphenyl)-n-hexane, meso-Hexestrol, Phenol, 4,4'-[(1R,2S)-1,2-diethyl-1,2-ethanediyl]bis-, rel-
IUPAC Name 4-[(3S,4R)-4-(4-hydroxyphenyl)hexan-3-yl]phenol
SMILES CC[C@H]([C@H](CC)c1ccc(O)cc1)c2ccc(O)cc2
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C18H22O2/c1-3-17(13-5-9-15(19)10-6-13)18(4-2)14-7-11-16(20)12-8-14/h5-12,17-20H,3-4H2,1-2H3/t17-,18+
Formula Weight 270.162
Percent Purity >95%
Chemical Name or Material Hexestrol
Grade HPLC