Learn More
Scharlab HEXAN PM 1L
Brand:  Scharlab HE02191000
HEXAN PM 1L
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 11314174
  • ONLINE EXCLUSIVE PRICE
    1128.75 NOK
    878.15 NOK / 1 L
    Save 250.60 NOK
    22% Off