Learn More
Hexamethyldisiloxane, Sigma-Aldrich™
Product Code. 15814281
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15814281 500 mL 500mL
15804281 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15814281 Supplier Sigma Aldrich Supplier No. 8140510500
 Restricted Item
Estimated Shipment: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 107-46-0
Specifications
Melting Point -68°C
Density 0.76 g/cm3 at 25°C
Boiling Point 100°C/1013 hPa
Molecular Formula ((CH3)3Si)2O
MDL Number MFCD00008265
Quantity 500 mL
Synonym Hexamethyldisiloxane, 2,2,4,4-Tetramethyl-3-oxa-2,4-disilapentane, HMDSO, Hexamethyldisiloxane
Molecular Weight (g/mol) 162.38
Grade Research