Learn More
Hexadecanoic-2,2-d2 Acid, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Hexadecanoic-2,2-D2 Acid |
| CAS | 62689-96-7 |
| Molecular Formula | C16 D2 H30 O2 |
| Quantity | 0.5 g |
| Synonym | Hexadecanoic-2,2-d2 acid (9 CI, ACI), 2,2-D2-Palmitic acid, 2,2-d2-Palmitic acid, Palmitic acid-2,2-2H2, α-Dideuterohexadecanoic acid, [2,2-2H2]Hexadecanoic acid, Hexadecanoic-2,2-D2 acid |
| InChI Formula | InChI=1 S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)/i15D2 |
| SMILES | [2 H]C([2 H])(CCCCCCCCCCCCCC)C(=O)O |
| IUPAC Name | 2,2-dideuteriohexadecanoic acid |
| Molecular Weight (g/mol) | 258.4364 |
| Formula Weight | 258.2528 g/mol |
| Show More |