missing translation for 'onlineSavingsMsg'
Learn More

Hesperin, MedChemExpress

Product Code. p-200099935
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16483338 10 mg 10mg
16456067 5 mg 5mg
16444586 100 mg 100mg
16439876 50 mg 50mg
4 options
This item is not returnable. View return policy

Product Code. 16483338

Brand: Medchem Express HY101371

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Hesperin is a bioactive ingredient present in Japanese horseradish (wasabi) and has been shown to be an Nrf2 activator.

Hesperin is a bioactive ingredient present in Japanese horseradish (wasabi) and has been shown to be an Nrf2 activator.

Specifications

Chemical Name or Material Hesperin
Physical Form Liquid
CAS 4430-35-7
Color Colorless
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C8H15NOS2
For Use With (Application) Cancer-programmed cell death
Solubility Information Ethanol : 100 mg/mL (487.00 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (243.50 mM; Need ultrasonic)
SMILES O=S(CCCCCCN=C=S)C
Molecular Weight (g/mol) 205.34
Formula Weight 205.34
Percent Purity 98.12%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.