missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HERC4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13691-1-AP-150UL
This item is not returnable.
View return policy
Description
Probable E3 ubiquitin-protein ligase involved in either protein trafficking or in the distribution of cellular structures. Required for spermatozoon maturation and fertility, and for the removal of the cytoplasmic droplet of the spermatozoon. E3 ubiquitin-protein ligases accept ubiquitin from an E2 ubiquitin- conjugating enzyme in the form of a thioester and then directly transfer it to targeted substrates.Specifications
| HERC4 | |
| Polyclonal | |
| Unconjugated | |
| HERC4 | |
| hect domain and RLD 4, HERC4, KIAA1593 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26091, 309758, 67345 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5GLZ8, Q5PQN1, Q6PAV2 | |
| Herc4 | |
| HERC4 Fusion Protein Ag4566 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction