Learn More
Thermo Scientific™ Heratherm™ Advanced Protocol Security Ovens
Product Code. 10307539 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
61 L
62 L
97 L
99 L
168 L
170 L
Type:
Gravity Convection
Mechanical Convection Oven
Electrical Requirements:
208/240 V 60 Hz
230 V 50/60 Hz
Material:
Coated Steel
Stainless Steel
Unit Size:
Each
Product Code. Capacity (Metric) Type Electrical Requirements Material unitSize
10307539 168 L Gravity Convection 230 V 50/60 Hz Coated Steel Each
11636430 61 L Gravity Convection 230 V 50/60 Hz Coated Steel Each
15324922 99 L Gravity Convection 208/240 V 60 Hz Coated Steel Each
10100020 99 L Gravity Convection 230 V 50/60 Hz Coated Steel Each
10222320 61 L Gravity Convection 230 V 50/60 Hz Stainless Steel Each
10232320 99 L Gravity Convection 230 V 50/60 Hz Stainless Steel Each
10110080 168 L Gravity Convection 230 V 50/60 Hz Stainless Steel Each
10100550 62 L Mechanical Convection Oven 230 V 50/60 Hz Coated Steel Each
12858231 97 L Mechanical Convection Oven 208/240 V 60 Hz Coated Steel Each
10820181 97 L Mechanical Convection Oven 230 V 50/60 Hz Coated Steel Each
11646440 170 L Mechanical Convection Oven 230 V 50/60 Hz Coated Steel Each
10660421 62 L Mechanical Convection Oven 230 V 50/60 Hz Stainless Steel Each
10672341 97 L Mechanical Convection Oven 230 V 50/60 Hz Stainless Steel Each
10775182 170 L Mechanical Convection Oven 230 V 50/60 Hz Stainless Steel Each
14 options
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 10307539
Brand: Thermo Scientific™ 51028147
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Ensure optimal sample protection with these advanced protocol security ovens that feature an extra layer of security for applications where process reliability and security are paramount.
Protect samples effectively with Thermo Scientific™ Heratherm™ Advanced Protocol Security Ovens, which feature an extra layer of security for applications where process reliability and security are paramount. A standard over-temperature alarm and an additional under-temperature alarm ensure sample stability. A door lock prevents disruption or tampering of samples, while a door alarm notifies the operator when the door is left open accidentally. Choose a stainless steel exterior for demanding pharmaceutical and clinical applications.
Heratherm Advanced Protocol Security Ovens are offered in a choice of gravity or mechanical convection. Gravity convection units are designed to protect delicate samples while offering temperature uniformity of ±2.5°C or better, and temperature stability of at least ±0.3°C at 150°C. Mechanical convection models provide temperature uniformity of ±1.5°C or better, and temperature stability of at least ±0.2°C at 150°C.
  • Broad temperatures range—up to 330°C
  • Optional sample sensor provides exact temperature of sample on display
  • Auto-dry function deactivates oven when the samples are dry, saving energy (Note: Optional sample sensor is required to utilize this feature)
  • Mechanical convection models: fan speed adjustable for application flexibility
  • Advanced digital timer for daily or weekly on / off cycles
  • Stainless-steel interior is easy to clean and corrosion resistant (steel quality AISI 304)
  • Programmable controller for temperature ramps and dwells
  • Up to 10 programs can be saved, each with 10 discrete steps
  • Electronically controlled fan speed and damper position can be incorporated
  • Programs can be repeated automatically
  • Access port allows the introduction of sensors for independent data monitoring
  • Simple calibration routine ensures temperature accuracy over time
  • Boost function enables rapid heating up—no need to operate the oven 24/7
Ordering Information:
Availability may vary by country.
TRUSTED_SUSTAINABILITY
Specifications
Controller Type Programmable Microprocessor control with vacuum fluorescent display
Capacity (Metric) 168 L
Temperature (Metric) 50°C to 330°C
Type Gravity Convection
Material Coated Steel
No. of Shelves 2 supplied/19 max.
Plug Type Country-specific
Temperature Stability ±0.3°C
Timer Weekly / Real time / Hour
Dimensions (D x W x H) Interior 22.2 x 17.2 x 26.8 in. (56.4 x 43.8 x 68 cm)
Dimensions (L x W x H) 29.1 x 25.2 x 36.2 in. (73.8 x 64 x 92 cm)
Length (English) 29.1 in.
Length (Metric) 73.8 cm
Width (English) 25.2 in.
Width (English) Interior 17.2 in.
Width (Metric) 64 cm
Width (Metric) Interior 43.8 cm
Height (English) 36.2 in.
Height (English) Interior 26.8 in.
Height (Metric) 92 cm
Height (Metric) Interior 68 cm
Depth (English) Interior 22.2 in.
Depth (Metric) Interior 56.4 cm
Electrical Requirements 230 V 50/60 Hz
Voltage 230 V
Wattage 3100 w
Amperage 13.5 A
Frequency 50/60 Hz
Show More Show Less
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.