Learn More
Thermo Scientific™ Herasafe™ KS Options and Accessories
Product Code. 10429183 Shop All Thermo Scientific Products
Click to view available options
Product Type:
1200 mm Indented Worksurface Module
150 mm Flat Worksurface Module
1500 mm Indented Worksurface Module
1800 mm Indented Worksurface Module
300 mm Flat Worksurface Module
300 mm Vubration Damped Worksurface
600 mm Flat Worksurface Module
600 mm Indented Worksurface Module
600 mm Vubration Damped Worksurface
900 mm Flat Worksurface Module
900 mm Indented Worksurface Module
KS12/KS15 Thimble Duct Exhaust Acc.
KS18 Thimble Duct Exhaust Acc.
KS9 Thimble Duct Exhaust Acc.
Utility Basket
Utility Basket with Hanging Bar
Unit Size:
Each
Product Code. 10429183
Brand: Thermo Scientific™ 50073770
Worksurfaces, exhaust transitions and baskets
TRUSTED_SUSTAINABILITY
Specifications
Description Thimble Duct Exhaust Transition for Herasafe KS 12 (4 ft) or KS 15 (5 ft) Cabinet
Product Type KS12/KS15 Thimble Duct Exhaust Acc.
For Use With (Equipment) Biosafety Cabinet