Learn More
Thermo Scientific™ Heraguard™ ECO Clean Bench UV Option
Integrate Thermo Scientific™ Heraguard™ ECO Clean Bench Accessories with the ECO clean bench to increase the flexibility and enhance operations.
Brand:  Thermo Scientific™ 51901073
Product Code. 15148912
  • ONLINE EXCLUSIVE PRICE
    8845.00 NOK
    8224.24 NOK / Each
    Save 620.76 NOK
    7% Off
Description
Integrate Thermo Scientific™ Heraguard™ ECO Clean Bench Accessories with the ECO clean bench to increase the flexibility and enhance operations. Options include media valves, solenoid valve, stainless-steel work surface wraps, side window modifications, UV and UV night covers, windscreen protection kits and additional accessories.
†Due to DC motors, Heraguard ECO clean benches use less energy to operate so there is less heat emitted into the lab than traditional AC motor clean benches. Based on data from internal testing and The University of Michigan Field Study, published in American Biotechnology Laboratory.