Learn More
Thermo Scientific™ Biological Safety Cabinets and Clean Bench Floor Stands
Product Code. 10171813 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Castors
Electronic Adjustable Floor Stand
Floor Stand
Foot Rest Bar
Height Adjustable Floor Stand
For Use With (Equipment):
Herasafe 2030i 0.9, Herasafe 2025 0.9, MSC Advantage 0.9
Herasafe 2030i 0.9, Herasafe 2025 0.9, MSC Advantage 0.9, Heraguard ECO 0.9
Herasafe 2030i 1.2, Herasafe 2025 1.2, MSC Advantage 1.2
Herasafe 2030i 1.2, Herasafe 2025 1.2, MSC Advantage 1.2, Heraguard ECO 1.2
Herasafe 2030i 1.5, Herasafe 2025 1.5, MSC Advantage 1.5
Herasafe 2030i 1.5, Herasafe 2025 1.5, MSC Advantage 1.5, Heraguard ECO 1.5
Herasafe 2030i 1.8, Herasafe 2025 1.8, MSC Advantage 1.8
Herasafe 2030i 1.8, Herasafe 2025 1.8, MSC Advantage 1.8, Heraguard ECO 1.8, 1.8/95
Herasafe 2030i, Herasafe 2025, MSC Advantage, Heraguard ECO
MSC Advantage 0.9
MSC Advantage 1.2
MSC Advantage 1.5
MSC Advantage 1.8
Unit Size:
Each
Pack of 4
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type For Use With (Equipment) Unit Size
10171813 Height Adjustable Floor Stand MSC Advantage 1.2 Each
10294014 Height Adjustable Floor Stand MSC Advantage 1.5 Each
10244394 Foot Rest Bar Herasafe 2030i 0.9, Herasafe 2025 0.9, MSC Advantage 0.9, Heraguard ECO 0.9 Each
11539915 Foot Rest Bar Herasafe 2030i 1.8, Herasafe 2025 1.8, MSC Advantage 1.8, Heraguard ECO 1.8, 1.8/95 Each
10546074 Electronic Adjustable Floor Stand Herasafe 2030i 1.5, Herasafe 2025 1.5, MSC Advantage 1.5 Each
10470484 Floor Stand MSC Advantage 0.9 Each
10060514 Electronic Adjustable Floor Stand Herasafe 2030i 0.9, Herasafe 2025 0.9, MSC Advantage 0.9 Each
10019882 Floor Stand MSC Advantage 1.8 Each
10690394 Height Adjustable Floor Stand MSC Advantage 1.8 Each
11509935 Castors Herasafe 2030i, Herasafe 2025, MSC Advantage, Heraguard ECO Pack of 4
11529915 Foot Rest Bar Herasafe 2030i 1.2, Herasafe 2025 1.2, MSC Advantage 1.2, Heraguard ECO 1.2 Each
10621154 Floor Stand MSC Advantage 1.2 Each
10101293 Electronic Adjustable Floor Stand Herasafe 2030i 1.2, Herasafe 2025 1.2, MSC Advantage 1.2 Each
10703605 Foot Rest Bar Herasafe 2030i 1.5, Herasafe 2025 1.5, MSC Advantage 1.5, Heraguard ECO 1.5 Each
10706255 Floor Stand MSC Advantage 1.5 Each
10090163 Electronic Adjustable Floor Stand Herasafe 2030i 1.8, Herasafe 2025 1.8, MSC Advantage 1.8 Each
10603955 Height Adjustable Floor Stand MSC Advantage 0.9 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Product Code. 10171813 Supplier Thermo Scientific™ Supplier No. 50109312
  • ONLINE EXCLUSIVE PRICE
    15055.00 NOK
    14756.01 NOK / Pack of 1
    Save 298.99 NOK
    1% Off
 Restricted Item
Estimated Shipment: 18-02-2027
Fixed, manual and electrically adjustable floor stands for the Thermo Scientific™ Clean Benches and Biological Safety Cabinets.
Choose from fixed, manual and electrically adjustable floor stands for the Thermo Scientific™ Clean Benches and Biological Safety Cabinets
†Due to DC motors, Heraguard ECO clean benches use less energy to operate so there is less heat emitted into the lab than traditional AC motor clean benches. Based on data from internal testing and The University of Michigan Field Study, published in American Biotechnology Laboratory.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) MSC Advantage 1.2
Description Provides variable ECO work height of 760 mm - 960 mm in 50 mm adjustable increments.
Product Type Height Adjustable Floor Stand
Height Overall (Metric) 680 to 880 mm
Working Height 750 to 950 mm