Learn More
Thermo Scientific™ Heraguard™ ECO Clean Bench Floor Stands
Shop All Thermo Scientific Products
Click to view available options
Product Type:
Adjustable Floor Stand
Castors
Electronic Adjustable Floor Stand
Floor Stand
Foot Rest Bar
For Use With (Equipment):
Heraguard ECO 0.9
Heraguard ECO 0.9, 1.2, 1.5, 1.8
Heraguard ECO 1.2
Heraguard ECO 1.5
Heraguard ECO 1.8, 1.8/95
Heraguard ECO 1.8, 1.8/95; Herasafe 2030i 1.8
Unit Size:
Each
Pack of 4
Description
Choose from fixed, manual and electrically adjustable Thermo Scientific™ Heraguard™ ECO Clean Bench Floors Stands for the Heraguard™ ECO clean bench.
†Due to DC motors, Heraguard ECO clean benches use less energy to operate so there is less heat emitted into the lab than traditional AC motor clean benches. Based on data from internal testing and The University of Michigan Field Study, published in American Biotechnology Laboratory.
Spécification
Spécification
| For Use With (Equipment) | Heraguard ECO 1.8, 1.8/95 |
| Description | Heraguard ECO Work Height of 760 mm to 960 mm plus Three Memory-set Heights |
| Product Type | Electronic Adjustable Floor Stand |
| Height Overall (Metric) | 680 to 880 mm |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu