Learn More
Thermo Scientific™ Heracell™ Gas Tight Inner Door
Shop All Thermo Scientific Products
Click to view available options
Material:
Glass
Unit Size:
Each
Description
6 gas tight doors replaces single inner door configuration
Specifications
Specifications
| Description | Gsa tight interior glass doors |
| For Use With (Equipment) | Heracell VIOS 250i CO2 incubator |
| Lighting | LED |
| Material | Glass |
| Type | Inner Door |
| Includes | 6 doors |