Learn More
Thermo Scientific™ Heracell™VIOS 250i CO2 Incubator Strengthened Stainless Steel Shelves
Shop All Thermo Scientific ProductsDescription
- 3 solid stainless steel shelves
- Includes 2 support rails
- Factory fitted
Specifications
Specifications
| Description | 3 stainless steel shelves |
| For Use With (Equipment) | Heracell VIOS 250i CO2 Incubators |
| Material | Stainless Steel |
| Type | Shelf |
| Includes | 3 shelves and 2 support rails |