missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ HepExtend™ Supplement (50X)

Product Code. 15661297
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15661297 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15661297 Supplier Gibco™ Supplier No. A2737501

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

HepExtend™ Supplement (50X) has been optimized for use with cryopreserved primary hepatocytes to improve cell viability, function, and number of days in culture. This enables researchers to perform metabolic and toxicity experiments not achievable using standard culture conditions.

HepExtend™ Supplement (50X) enables researchers to perform metabolic and toxicity experiments not achievable using standard culture conditions. Simply add HepExtend™ Supplement to standard William’s E Hepatocyte Maintenance Media and use with your current hepatocyte culturing methods and instrumentation. The supplement does not contain any small molecules or fetal bovine serum, components that are known to interfere with primary hepatocyte function.

  • Enabling—designed to provide maximum cell viability and cell health when culturing primary plateable hepatocytes
  • Extended cell survival—days in culture can be extended to >10 days while maintaining normal cell morphology and bile canaliculi
  • Consistent—manufactured in a cGMP-compliant facility using the highest quality materials

Maintain P450 activity

HepExtend™ Supplement can be used with your standard hepatocyte culture media to help facilitate metabolically competent hepatocytes for at least 10 days for use in downstream ADME/Tox applications not achievable with standard 5-day culture systems.

Enhanced gene expression

HepExtend™ supplementation increases the basal gene expression of many key genes involved in drug metabolism and transport.

Retain inducibility

Nuclear receptor activation and gene inducibility is retained in cultures treated with HepExtend™ supplemented media.

TRUSTED_SUSTAINABILITY

Specifications

Classification Animal Origin-free
Culture Type Hepatocyte Cell Culture
Form Liquid
Product Type Hepatocyte Supplement
pH 6 to 8
Product Line HepExtend
Quantity 10 mL
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.