Learn More
HEPES (Fine White Crystals/Molecular Biology), Fisher BioReagents™
Product Code. 10081113
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Glass Bottle
Poly Bottle
Poly Pail
Unit Size:
100g
1kg
500g
5kg
Product Code. Quantity Packaging unitSize
10081113 1 kg Poly Bottle 1kg
10397023 100 g Glass Bottle 100g
10756254 500 g Poly Bottle 500g
10619163 5 kg Poly Pail 5kg
4 options
Product Code. 10081113
Brand: Fisher BioReagents BP3101
Request Bulk or Custom Format
Commonly used buffering agent | CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol
Commonly used buffering agent
HEPES is commonly used as a buffering agent.
TRUSTED_SUSTAINABILITY
Specifications
Absorbance 0.01 max. (0.1M solution) at 280nm
Chemical Name or Material HEPES
CAS 7365-45-9
Color White
pH 5.0 to 6.5
Physical Form Powder or Solid
Quantity 1 kg
DNase DNase free
Purity Grade Notes DNase-, RNase- and Protease-Free
Assay Percent Range ≥99 %
Identification Pass Test
Molecular Formula C8H18N2O4S
MDL Number MFCD00006158
Packaging Poly Bottle
Protease Protease free
Merck Index 15, 4689
Synonym N-(2-Hydroxyethyl)piperazine-N«-2-ethanesulfonic Acid
Solubility Information Soluble in water
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
Molecular Weight (g/mol) 238.30
PubChem CID 23831
ChEBI CHEBI:42334
Percent Purity ≥99%
Grade Molecular Biology