Learn More
Karl Hecht™ Heparinised Micro Hematocrit Capillaries
Product Code. 10460742
Change view
Click to view available options
Quantity:
100 Pieces/Tube; 10 Tubes/Box
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10460742 100 Pieces/Tube; 10 Tubes/Box Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Product Code. 10460742 Supplier Karl Hecht™ Supplier No. 40565010
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
A fire-polished hematocrit capillary with sodium heparin.
  • Disposable
  • With mark at 60mm
  • With red color code
  • Fire-polished at color-coded end
  • Total capacity of approx. 18μL
  • Capacity up to mark approx. 14μL
  • With CE marking
  • 100 pieces/tube; 10 tubes/box
Specifications
Capacity (Metric) 18 μL, 14 μL to mark
Diameter (Metric) Inner 0.55 mm
Quantity 100 Pieces/Tube; 10 Tubes/Box
Length (Metric) 75 mm
Certifications/Compliance CE