Learn More
Hemopexin Mouse anti-Human, Porcine, Rat, Clone: 3A9D6, Proteintech
Product Code. 16828395 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16828395 20 μL 20µL
16818395 150 μL 150µL
2 options
Product Code. 16828395
Brand: Proteintech 664791IG20UL
Mouse Monoclonal Antibody
Hemopexin is a serum glycoprotein that binds heme and transports it to the liver for breakdown and iron recovery, after which the free hemopexin returns to the circulation. Structurally, hemopexin consists of two similar halves of approximately two hundred amino acid residues connected by a histidine-rich hinge region. Each half is itself formed by the repetition of a basic unit of some 35 to 45 residues.
Specifications
Antigen Hemopexin
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3A9D6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene HPX
Gene Accession No. P02790, P20059, P50828
Gene Alias Beta 1B glycoprotein, hemopexin, HPX
Gene Symbols HPX
Host Species Mouse
Immunogen Hemopexin Fusion Protein Ag8533
Purification Method Protein G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3263, 396998, 58917
Target Species Pig, Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1