Learn More
Hematoxylin Trihydrate, Fisher BioReagents
Product Code. 10451905
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Amber Glass
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10451905 25 g Amber Glass 25g
10012694 5 g Amber Glass 5g
10072594 100 g Amber Glass 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10451905 Supplier Fisher BioReagents Supplier No. BP252325
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Hematoxylin Trihydrate is used mainly as a stain in microscopy.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 517-28-2
Molecular Formula C16H14O6
Molecular Weight (g/mol) 302.28
MDL Number MFCD00078111
InChI Key WZUVPPKBWHMQCE-WKTCHCBJNA-N
Synonym Natural Black I
PubChem CID 45029742
SMILES [H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3
Specifications
Melting Point 100°C
Merck Index 15, 4672
Identification Pass Test
IUPAC Name (1R,10S)-8-oxatetracyclo[8.7.0.0²,⁷.0¹²,¹⁷]heptadeca-2(7),3,5,12(17),13,15-hexaene-5,6,10,14,15-pentol
Lambda Max 290 to 292nm
Formula Weight 356.4
Loss on Drying Report %
Packaging Amber Glass
Color Amber
Quantity 25 g
Physical Form Powder
Chemical Name or Material Hematoxylin Trihydrate