Learn More
Hematoxylin hydrate, 96% (dry wt.), water ca 6%
Product Code. p-7232723
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. 11499633
Brand: Thermo Scientific Alfa Aesar A12431.06
Request Bulk or Custom Format
CAS: 517-28-2 | C{1}{6}H{1}{4}O{6}^.xH{2}O
Extracted from the bark of the logwood tree
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Extracted from the bark of the logwood tree
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 517-28-2
Molecular Formula C{1}{6}H{1}{4}O{6}^.xH{2}O
Synonym C.I. 75290;Natural Black 1
Specifications
Melting Point ca 200°C (decomposition)
Quantity 5 g
Percent Purity 96%
compliance-icons
Product Identifier
  • Hematoxylin hydrate, water ca 6%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Hematoxylin hydrate