Learn More
Greiner Bio-One Hematology K2 EDTA Evacuated Tubes
Product Code. 13018672
Change view
Click to view available options
Capacity (Metric):
4 mL
Closure Color:
Lavender
Unit Size:
Pack of 1200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Color Unit Size
13018672 4 mL Lavender Pack of 1200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13018672 Supplier Greiner Bio-One Supplier No. 454209
 Restricted Item
Estimated Shipment: 19-10-2026
For routine immunohematology testing
  • Tube material: Polyethylene therephthalate
  • Cap color/ring color: lavender/black/yellow
Compliance
CE Marked
TRUSTED_SUSTAINABILITY
Specifications
Additive K2 EDTA
Material PET
Closure Type Safety Pull Cap
Capacity (Metric) 4 mL
Closure Color Lavender
For Use With (Application) For routine immunohematology testing
Diameter (Metric) Outer 13 x 75 mm