Learn More
Hirschmann™ Hematocrit Tubes
Product Code. 30090389
Change view
Click to view available options
Capacity (Metric):
60 μL
75 μL
Unit Size:
Pack of 1000
Pack of 5000
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
30090389 75 μL Pack of 5000
10146260 60 μL Pack of 1000
30090386 60 μL Pack of 5000
10604162 75 μL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30090389 Supplier Hirschmann™ Supplier No. 9100175
  • ONLINE EXCLUSIVE PRICE
    1850.00 NOK
    1626.13 NOK / Pack of 5000
    Save 223.87 NOK
    12% Off
View all promotions
 Restricted Item
Estimated Shipment: 17-12-2026
Excellent for centrifuging blood in the laboratory. Hirschmann Instruments™ Hematocrit Tubes are manufactured from disposable capillary glass.
  • Not heparinized
  • Color-coded blue
  • Acc. to DIN EN ISO 12772, CE acc. IVD 98/79 EC
Specifications
Product Type Hematocrit Tube
Capacity (Metric) 75 μL
Diameter (Metric) Inner 1.07/1.23 mm
For Use With (Application) For centrifuging blood samples, not heparinized
Length (Metric) 75 ±0.5 mm
Certifications/Compliance DIN EN ISO 12772, CE, IVD 98/79 EC