Learn More
Hirschmann™ Hematocrit Tubes
Description
- Not heparinized
- Color-coded blue
- Acc. to DIN EN ISO 12772, CE acc. IVD 98/79 EC
Specifications
Specifications
| Product Type | Hematocrit Tube |
| Capacity (Metric) | 60 μL |
| Diameter (Metric) Inner | 0.9/1.0 mm |
| For Use With (Application) | For centrifuging blood samples, not heparinized |
| Length (Metric) | 75 ±0.5 mm |
| Certifications/Compliance | DIN EN ISO 12772, CE, IVD 98/79 EC |