Learn More
Hirschmann™ Sodium Heparinized Hematocrit Tubes
Product Code. 30090390
Change view
Click to view available options
Capacity (Metric):
60 μL
75 μL
100 μL
Unit Size:
Pack of 1000
Pack of 5000
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
30090390 100 μL Pack of 5000
10233391 100 μL Pack of 1000
10410772 60 μL Pack of 1000
30090388 60 μL Pack of 5000
10199641 75 μL Pack of 1000
30090387 75 μL Pack of 5000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 30090390 Supplier Hirschmann™ Supplier No. 9100299
  • ONLINE EXCLUSIVE PRICE
    3590.00 NOK
    3006.59 NOK / Pack of 5000
    Save 583.41 NOK
    16% Off
View all promotions
 Restricted Item
Estimated Shipment: 08-02-2027
Excellent for centrifuging blood in the laboratory. Hirschmann Instruments™ Sodium Heparinized Hematocrit Tubes are manufactured from disposable capillary glass.
  • Color-coded red
  • Acc. to DIN EN ISO 12772, CE acc. IVD 98/79 EC
Specifications
Product Type Hematocrit Tube Na-hep
Capacity (Metric) 100 μL
For Use With (Application) For centrifuging blood samples, Sodium heparinized
Certifications/Compliance DIN EN ISO 12772, CE, IVD 98/79 EC