Learn More
Hirschmann™ Sodium Heparinized Hematocrit Tubes
Product Code. 10199641
Change view
Click to view available options
Capacity (Metric):
60 μL
75 μL
100 μL
Unit Size:
Pack of 1000
Pack of 5000
Product Code. Capacity (Metric) unitSize
10199641 75 μL Pack of 1000
10233391 100 μL Pack of 1000
30090390 100 μL Pack of 5000
10410772 60 μL Pack of 1000
30090388 60 μL Pack of 5000
30090387 75 μL Pack of 5000
6 options
Product Code. 10199641
Brand: Hirschmann™ 9100275
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Excellent for centrifuging blood in the laboratory. Hirschmann Instruments™ Sodium Heparinized Hematocrit Tubes are manufactured from disposable capillary glass.
  • Color-coded red
  • Acc. to DIN EN ISO 12772, CE acc. IVD 98/79 EC
Specifications
Product Type Hematocrit Tube Na-hep
Capacity (Metric) 75 μL
Diameter (Metric) Inner 1.07/1.23 mm
For Use With (Application) For centrifuging blood samples, Sodium heparinized
Length (Metric) 75 ±0.5 mm
Certifications/Compliance DIN EN ISO 12772, CE, IVD 98/79 EC