Learn More
Helios Italquartz Quartz Crucibles
Description
- Chemically inert to acids (except HF)
- Available in a wide range of volumes
- Available in transparent or translucent color
- Available in tall or short form
- Resistant to corrosion
Specifications
Specifications
| Color | Transparent |
| Description | Short form |
| Material | Quartz |
| Max. Temperature (Metric) | 1000°C |
| Capacity (Metric) | 75 mL |
| Diameter (Metric) Outer | 67mm |
| Height (Metric) | 45mm |
| Product Type | Crucible |