Learn More
Helios Italquartz Quartz Crucibles
Product Code. 11735694
Click to view available options
Capacity (Metric):
15 mL
20 mL
25 mL
30 mL
50 mL
75 mL
130 mL
190 mL
Diameter (Metric) Outer:
35mm
41mm
43mm
47mm
50mm
57mm
67mm
75mm
80mm
Color:
Translucent
Transparent
Description:
Short form
Tall form
Unit Size:
Each
Product Code. 11735694
Brand: Helios Italquartz 63F00513
Only null left
Add to basket
Add to basket
Excellent for storage of samples in the laboratory. Helios Italquartz™ Quartz Crucibles offer good thermal shock resistance up to 1000°C.
  • Chemically inert to acids (except HF)
  • Available in a wide range of volumes
  • Available in transparent or translucent color
  • Available in tall or short form
  • Resistant to corrosion
Specifications
Color Translucent
Description Tall form
Material Quartz
Max. Temperature (Metric) 1000°C
Capacity (Metric) 50 mL
Diameter (Metric) Outer 50mm
Height (Metric) 50mm
Product Type Crucible