Learn More
Helios™ Helios Italquartz™ Evaporating Dishes
Product Code. p-8011805
Change view
Click to view available options
Capacity (Metric):
20 mL
55 mL
Diameter (Metric):
47 mm
61 mm
77 mm
Height (Metric):
15 mm
18 mm
27 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Diameter (Metric) Height (Metric) unitSize
11734056 20 mL 47 mm 15 mm Each
11700645 55 mL 61 mm 27 mm Each
11724056 55 mL 77 mm 18 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11734056 Supplier Helios™ Supplier No. 61F00146
Specifications
Bottom Shape Flat
Capacity (Metric) 20 mL
Material Quartz
Shape Round
Type Evaporating Dish
Height (Metric) 15 mm
Diameter (Metric) 47 mm